SY3H5815FB2B Synthonix Corporation

(4-((Tetrahydro-2H-pyran-2-yl)oxy)phenyl)boronic acid

  • Cas Number : 182281-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/3812.html

Safety Information

MSDS https://synthonix.com/3812.html
Availability details for
Quantity
  • Availability
  • Add to Cart