SY3H5815FB3A Synthonix Corporation

3-(Trifluoromethyl)-1H-pyrazole-4-carbaldehyde

  • Cas Number : 1001020-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/9855.html

Safety Information

MSDS https://synthonix.com/9855.html
Availability details for
Quantity
  • Availability
  • Add to Cart