SY3H5815FB3E Synthonix Corporation

1,4-Oxazepan-5-one

  • Cas Number : 10341-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/16165.html

Safety Information

MSDS https://synthonix.com/16165.html
Availability details for
Quantity
  • Availability
  • Add to Cart