SY3H5815FB3F Synthonix Corporation

2-oxo-2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine-6-carbonitrile

  • Cas Number : 1190317-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17170.html

Safety Information

MSDS https://synthonix.com/17170.html
Availability details for
Quantity
  • Availability
  • Add to Cart