SY3H5815FB41 Synthonix Corporation

1-Oxa-7-azaspiro[3.5]nonane oxalate

  • Cas Number : 1408076-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/19484.html

Safety Information

MSDS https://synthonix.com/19484.html
Availability details for
Quantity
  • Availability
  • Add to Cart