SY3H5815FB43 Synthonix Corporation

2-Oxa-7-azaspiro[4.5]decane hydrochloride

  • Cas Number : 374795-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23357.html

Safety Information

MSDS https://synthonix.com/23357.html
Availability details for
Quantity
  • Availability
  • Add to Cart