SY3H5815FB44 Synthonix Corporation

Oxazole-2-carboxamide

  • Cas Number : 884539-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26086.html

Safety Information

MSDS https://synthonix.com/26086.html
Availability details for
Quantity
  • Availability
  • Add to Cart