SY3H6E40F016 Synthonix Corporation

tert-butyl 5-(hydroxymethyl)-2-azabicyclo[2.2.1]heptane-2-carboxylate

  • MDL number NO MDL FOUND
  • Cas Number : 1363210-35-8
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 5-(hydroxymethyl)-2-azabicyclo[2.2.1]heptane-2-carboxylate
Inchi Code 1S/C12H21NO3/c1-12(2,3)16-11(15)13-6-8-4-10(13)5-9(8)7-14/h8-10,14H,4-7H2,1-3H3
Inchi Key FZSNTGVWRIIJNL-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/19046.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/19046.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart