SY3H6E4130AA Synthonix Corporation

1-Ethynylcyclopropan-1-amine hydrochloride

  • MDL number MFCD28404680
  • Cas Number : 1268810-17-8
  • Molecular Weight : 117.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-ethynylcyclopropan-1-amine hydrochloride
Inchi Code 1S/C5H7N.ClH/c1-2-5(6)3-4-5;/h1H,3-4,6H2;1H
Inchi Key XBHDGYBZTODLSE-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/64486.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/64486.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart