SY3H6E4131A0 Synthonix Corporation

Ethyl 3-(4-(methylamino)-3-nitro-N-(pyridin-2-yl)benzamido)propanoate

  • MDL number MFCD09833623
  • Cas Number : 429659-01-8
  • Molecular Weight : 372.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/66549.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/66549.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart