SY3H6E41480B Synthonix Corporation

4-(1H-Imidazol-1-yl)piperidine dihydrochloride

  • Cas Number : 403492-40-0
  • Molecular Weight : 224.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/60933.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/60933.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart