SY3H6E414853 Synthonix Corporation

3-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number NO MDL FOUND
  • Cas Number : 1627924-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/67114.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/67114.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart