SY3H7CF2AF52 Synthonix Corporation

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-cyanophenyl)propanoic acid

  • MDL number MFCD01863771
  • Cas Number : 401933-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/62309.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/62309.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart