SY3H7CF2B0AA Synthonix Corporation

rel-tert-Butyl (3aR,6aS)-5-(hydroxymethyl)hexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate

  • MDL number NO MDL FOUND
  • Cas Number : 2220998-56-9
  • Molecular Weight : 241.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (3aR,5s,6aS)-5-(hydroxymethyl)hexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate
Inchi Code 1S/C13H23NO3/c1-13(2,3)17-12(16)14-6-10-4-9(8-15)5-11(10)7-14/h9-11,15H,4-8H2,1-3H3/t9-,10+,11-
Inchi Key XRNJMGKDWQLIJX-JGPRNRPPSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/69833.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/69833.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart