SY3H93DAEF4F Synthonix Corporation

trans-2-((5-Bromo-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-indazol-3-yl)amino)-1-((1r,4r)-4-((tert-butyldimethylsilyl)oxy)cyclohexyl)-1H-benzo[d]imidazole-5-carboxylic acid

  • MDL number NO MDL FOUND
  • Cas Number : 1415565-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/65955.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/65955.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart