SY3H93DAEF52 Synthonix Corporation

tert-butyl 4-(4-bromo-5-methyl-pyrazol-1-yl)piperidine-1-carboxylate

  • MDL number NO MDL FOUND
  • Cas Number : 1076223-99-8
  • Molecular Weight : 344.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-(4-bromo-5-methyl-1H-pyrazol-1-yl)piperidine-1-carboxylate
Inchi Code 1S/C14H22BrN3O2/c1-10-12(15)9-16-18(10)11-5-7-17(8-6-11)13(19)20-14(2,3)4/h9,11H,5-8H2,1-4H3
Inchi Key CMZCIVZMQCWOEK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/69913.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/69913.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart