SY3H93DAEF57 Synthonix Corporation

(4aS,6aR,6bR,10S,12aR,14aR,14bR)-methyl 10-acetoxy-2,2,6a,6b,9,9,12a-heptamethyl-14-oxo-docosahydropicene-4a-carboxylate

  • MDL number NO MDL FOUND
  • Cas Number : 25493-94-1
  • Molecular Weight : 528.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl (4aS,6aR,6bR,10S,12aR,14aR,14bS)-10-acetoxy-2,2,6a,6b,9,9,12a-heptamethyl-14-oxoicosahydropicene-4a(2H)-carboxylate
Inchi Code 1S/C33H52O5/c1-20(34)38-25-11-12-30(6)23(29(25,4)5)10-13-31(7)24(30)18-22(35)26-21-19-28(2,3)14-16-33(21,27(36)37-9)17-15-32(26,31)8/h21,23-26H,10-19H2,1-9H3/t21-,23?,24?,25-,26-,30-,31+,32+,33-/m0/s1
Inchi Key WJXGKADQKHUESG-AFFXIZASSA-N

Description

Purity 95%
Storage temp. Refrigerator
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/65921.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/65921.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart