SY3H94E1CA9E Synthonix Corporation

(5-Bromopyridin-2-yl)zinc iodide, 0.50 M in THF

  • Cas Number : 1065486-64-7
  • Molecular Weight : 349.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/73108.html

Safety Information

signalword Danger
pcodes P222, P223, P231, P232, P261, P280
MSDS https://synthonix.com/73108.html
pictograms GHS02, GHS05
hstatements H225, H302, H314, H335, H351
Availability details for
Quantity
  • Availability
  • Add to Cart