TA9H93ED7229 Target Molecule Corp.

Isoastragaloside IV

  • Cas Number : 136033-55-1
  • Molecular Weight : 784.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98.00
Storage temp. -20
Physical Form Powder
Shipping Temperature 0�
Country of Origin US
Product Link www.targetmol.com/compound/Isoastragaloside IV

Safety Information

MSDS www.targetmol.com/compound/Isoastragaloside IV
Availability details for
Quantity
  • Availability
  • Add to Cart