EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Identification of sulfur-containing metabolites of 2,4,2',4'-tetrachlorobiphenyl in mice.
PMID 100159
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
283533
2,4-Dichloroiodobenzene, 98%
C6H3Cl2I