EMAIL THIS PAGE TO A FRIEND
Cell and tissue research
Tetrodotoxin suppresses morphological enhancement of the metastatic MAT-LyLu rat prostate cancer cell line.
PMID 10022970
Abstract
Voltage-gated Na+ channels are expressed by highly metastatic MAT-LyLu cells, but not by poorly metastatic AT-2 cells, derived from the rodent Dunning model of prostatic cancer. We have investigated the possible involvement of these channels in the morphological development of the cells. Incubation of both the MAT-LyLu and the AT-2 cell line for 24 h with the Na+ channel blocker tetrodotoxin (TTX) at 6 microM altered the morphology only of the MAT-LyLu cell line. TTX produced significant decreases in: (a) cell process length and (b) field diameter, and increases in (c) cell body diameter and (d) process thickness. Importantly, 6 microM TTX had no significant effects on proliferation rates or cellular toxicity. The results suggest that Na+ channel activity plays a significant role in determining the morphological development of MAT-LyLu cells in such a way as to enhance their metastatic potential.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
MAT-LyLu, 94101454