EMAIL THIS PAGE TO A FRIEND
Biophysical chemistry
Influence of 5-bromodeoxycytosine substitution on triplex DNA stability and conformation.
PMID 10028230
Abstract
Three triple-helical hairpin DNAs with substitution of 5-bromocytosine for cytosine in different strands have been investigated by molecular mechanics and Raman spectroscopy. The stability of the three substituted triplexes were compared with the corresponding unsubstituted triplex DNA by the molecular mechanics method. Base stacking interactions and strand--strand interactions of each triplex were analyzed in detail. Sugar conformations in these triplexes have been determined by both vibrational spectroscopy and molecular dynamics simulation. The hairpin triplexes with substitution occurring in strand I or both in strands I and III have the main sugar conformation of C3'-endo, while the triplex with substitution occurring in strand III is the combination of C3'-endo and C2'-endo sugar conformation. Theoretical results are basically in agreement with experiments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
150649
5-Bromocytosine, 99%
C4H4BrN3O