EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Purification of monoclonal antibodies from cell culture supernatants using a modified zirconia based cation-exchange support.
PMID 10028281
Abstract
A method suitable for the isolation of monoclonal antibodies (Mabs) is described. The protocol utilizes a zirconia based column modified with ethylenediamine-N,N'-tetra(methylenephosphonic) acid to create a novel cation-exchange chromatographic support. Initial experiments using a linear salt gradient demonstrate the ability of this support to efficiently separate Mab from transferrin and bovine serum albumin in a model matrix. Results of the purification of Mab from an actual cell culture supernatant over a range in protein concentrations are also shown. Analyses by enzyme-linked immunosorbent assay and gel electrophoresis demonstrate that Mabs can be recovered from a cell culture supernatant at high yield (92-98%) and high purity (> 95%) in a single chromatographic step.