EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
Metabolism of liriodendrin and syringin by human intestinal bacteria and their relation to in vitro cytotoxicity.
PMID 10071956
Abstract
When liriodendrin or syringin was incubated for 24 h with human intestinal bacteria, two metabolites, (+)-syringaresinol-beta-D-glucopyranoside and (+)-syringaresinol, from liriodendrin and one metabolite, synapyl alcohol, from syringin were produced. The metabolic time course of liriodendrin was as follows: at early time, liriodendrin was converted to (+)-syringaresinol-beta-D-glucopyranoside, and then (+)-syringaresinol. The in vitro cytotoxicities of these metabolites, (+)-syringaresinol and synapyl alcohol, were superior to those of liriodendrin and syringin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
08198
Eleutheroside E, analytical standard
C34H46O18
SMB00181
Liriodendrin, ≥85% (LC/MS-ELSD)
C34H46O18