EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
Biotransformation of theophylline in cirrhotic rats induced by biliary obstruction.
PMID 10071961
Abstract
The object of this work was to study the pharmacokinetic differences and the cause of these differences in cirrhotic rats induced by biliary obstruction when aminophylline (8 mg/kg as theophylline, i.v.) was administered. The concentrations of theophylline and its major metabolite (1,3-dimethyluric acid) in plasma were determined by HPLC. In addition, formation of 1,3-dimethyluric acid from theophylline in microsomes and the changes in the activity of drug metabolizing enzymes, which are suggested to be involved in theophylline metabolism, were determined. In cirrhotic rats, the systemic clearance of theophylline was reduced to 30% of the control value while AUC (area under the plasma concentration-time curve) and (t1/2)beta were increased 1.3 fold and 3.5 fold, respectively. The formation of 1,3-dimethyluric acid was decreased to 30% of the control value in microsomes of cirrhotic rat liver. In cirrhotic rat liver, activities of aniline hydroxylase (CYP2E1 related), erythromycin-N-demethylase (CYP3A related), and methoxyresorufin-O-demethylase (CYP1A2 related), which were reported to be related with theophylline metabolism, were decreased to 67%, 53%, and 76% that of normal rat liver, respectively. From the results, it can be concluded that in cirrhotic rats induced by biliary obstruction, the total body clearance of theophylline is markedly reduced and it may be due to decreased activity of drug metabolizing enzymes in liver.