EMAIL THIS PAGE TO A FRIEND
Journal of natural products
New cytotoxic oxygenated fucosterols from the brown alga Turbinaria conoides.
PMID 10075746
Abstract
Fucosterol (1), 24xi-hydroperoxy-24-vinylcholesterol (2), 29-hydroperoxystigmasta-5,24(28)-dien-3beta-ol (3), 24-ethylcholesta-4,24(28)-dien-3-one (4), 24xi-hydroperoxy-24-ethylcholesta-4,28(29)-dien-3-one (5), 24-ethylcholesta-4,24(28)-dien-3,6-dione (6), 24xi-hydroperoxy-24-ethylcholesta-4,28(29)-dien-3,6-di one (7), 6beta-hydroxy-24-ethylcholesta-4,24(28)-dien-3-one (8), and 24xi-hydroperoxy-6beta-hydroxy-24-ethylcholesta-4,28(2 9)-dien-3-one (9) were isolated from the marine brown alga Turbinaria conoides. The structures of these compounds were established by spectral analysis. Isolated for the first time from a natural source, the oxygenated fucosterols 4-9 exhibit cytotoxicity against various cancer cell lines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5379
Fucosterol, ≥93%
C29H48O