EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Phosphodiesterase 4-dependent regulation of cyclic AMP levels and leukotriene B4 biosynthesis in human polymorphonuclear leukocytes.
PMID 10079010
Abstract
Several selective phosphodiesterase 4 inhibitors were found to be potent inhibitors of the N-formyl-Met-Leu-Phe (fMLP)-induced leukotriene B4 biosynthesis by human polymorphonuclear leukocytes with IC50s in the nanomolar range (0.09-26 nM). The rank order of potency was 6-(4-pyridylmethyl)-8-(3-nitrophenyl)quinoline (RS-14203) > 3-benzyl-5-phenyl-3H-imidazo[4,5-c][1,8]naphthyridin-4(5H)-one (KF18280) > 8-aza-1-(3-nitrophenyl)-3-(4-pyridylmethyl)-2,4-quinazoline dione (RS-25344) > 3-cyclo-pentyloxy-N-[3,5-dichloro-4-pyridyl]-4-methoxybenzamide (RP-73401) > R-rolipram > R-4-[2-(3-cyclopentyloxy-4-methoxyphenyl)-2-phenylethyl] pyridine (CDP840)> S-rolipram. Isoproterenol (IC50 = 350 nM) and prostaglandin E2 (IC50 = 59 nM) also suppressed leukotriene B4 biosynthesis. Inhibitors of the phosphodiesterase 1 (8-methoxymethyl-1-methyl-3-(2-methylpropyl)xanthine (8-MeOMe-IBMX)), phosphodiesterase 2 (erythro-9-(2-hydroxy-3-nonyl)adenine (EHNA)), phosphodiesterase 3 (quazinone and milrinone) and phosphodiesterase 5 (zaprinast and dipyridamole) had no inhibitory effects on the fMLP-induced leukotriene B4 biosynthesis (IC50s > 20 microM). All phosphodiesterase 4 inhibitors caused an accumulation of cellular cyclic AMP to 140-185% over the basal level of fMLP-treated control cells, comparable to that observed with high concentrations of isoproterenol and prostaglandin E2. In contrast, the complete inhibition of leukotriene B4 production by 5-lipoxygenase and 5-lipoxygenase-activating protein (FLAP) inhibitors had no effect on cyclic AMP levels. Phosphodiesterase 1, 2, 3 and 5 inhibitors had little effect on the level of cellular cyclic AMP (89-126% of the basal cyclic AMP level). Dose-dependencies for R-rolipram, RS-14203 and CDP840 indicated that the maximal accumulation of cyclic AMP occurred at concentrations of phosphodiesterase 4 inhibitors higher than those required for the inhibition of leukotriene B4 production. The presence of a mixture of 8-MeOMe-IBMX, EHNA, milrinone and zaprinast to inhibit phosphodiesterase 1, 2, 3 and 5 had little effect on the dose-dependence of R-rolipram for the inhibition of leukotriene B4 biosynthesis or cyclic AMP accumulation. These data demonstrate that selective phosphodiesterase 4 inhibitors can inhibit the fMLP-induced leukotriene B4 biosynthesis in human polymorphonuclear leukocytes with a potency similar or greater than that of potent 5-lipoxygenase or FLAP inhibitors. This inhibition is accompanied by small variations in the levels of cellular cyclic AMP and appears to proceed independently of the other phosphodiesterases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9766
Dipyridamole, ≥98% (HPLC)
C24H40N8O4