EMAIL THIS PAGE TO A FRIEND
Neurologia (Barcelona, Spain)
[Acute familial dystonia caused by antidopaminergic drugs].
PMID 10079694
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8289
Clebopride maleate salt, ~95%
C20H24ClN3O2 · C4H4O4