EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Mutation spectrum of ATP7A, the gene defective in Menkes disease.
PMID 10079817
Abstract
Our knowledge about Menkes disease (MD) has expanded greatly since its description in 1962 as a new X-linked recessive neurodegenerative disorder of early infancy. Ten years later a defect in copper metabolism was established as the underlying biochemical deficiency. In the beginning of 1990s efforts were concentrated on the molecular genetic aspects. The disease locus was mapped to Xq13.3 and the gene has been isolated by means of positional cloning. This was the beginning of a series of new findings which have greatly enhanced our understanding of copper metabolism not only in human, but also in other species. This review will focus on the molecular genetic aspects of Menkes disease and its allelic form occipital horn syndrome. The mutations will be compared briefly with those described in the animal model mottled mouse, and in Wilson disease, the autosomal recessive disorder of copper metabolism.