EMAIL THIS PAGE TO A FRIEND
Nature genetics
Mutant NDUFV1 subunit of mitochondrial complex I causes leukodystrophy and myoclonic epilepsy.
PMID 10080174
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV48312
Anti-NDUFV1 antibody produced in rabbit, IgG fraction of antiserum