EMAIL THIS PAGE TO A FRIEND
Journal of human genetics
Chromosomal assignment of a human apoptosis-associated tyrosine kinase gene on chromosome 17q25.3 by somatic hybrid analysis and fluorescence in situ hybridization.
PMID 10083745
Abstract
Here, we report on the chromosomal location of the human apoptosis-associated tyrosine kinase gene. Based on polymerase chain reaction analysis with a human/rodent monochromosomal hybrid cell panel and fluorescence in situ hybridization, the gene was mapped on 25.3 region of chromosome 17.