EMAIL THIS PAGE TO A FRIEND
Pneumologie (Stuttgart, Germany)
[Pulmonary diagnosis with model aerosols. II: Aerosol bolus dispersion--a marker for convection gas transport. Basic principles and initial clinical results].
PMID 10091519