EMAIL THIS PAGE TO A FRIEND
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology
Selective enhancement of production of IgE, IgG4, and Th2-cell cytokine during the rebound phenomenon in atopic dermatitis and prevention by suplatast tosilate.
PMID 10094221
Abstract
Atopic dermatitis is a chronic inflammatory skin disease, which is commonly treated with topical steroids. It is, however, associated with rebound after therapy has been discontinued. This study was designed to elucidate the mechanisms of the rebound phenomenon, and to test the effect of an oral anti-allergic medication, suplatast tosilate, on atopic dermatitis. This is a randomized, placebo controlled study. Patients with atopic dermatitis who had been treated with strong steroid ointment (dexamethasone valerate) for several years were divided into two groups. One group (the control group, n = 15) was treated with a non-steroid anti-inflammatory ointment (bufexamac ointment), while the other group (the suplatast tosilate group, n = 17) was treated with the anti-allergic medications, suplatast tosilate and bufexamac ointment. In each group, in vitro production of immunoglobulins and cytokines before and after 2 weeks of treatment was measured. In the control group, 15 of the 15 patients experienced rebound and mean production of IgE, IgG4, IL-4, IL-5, IL-10, and IL-13 was enhanced after 2 weeks. In contrast, only 2 of the 17 patients in the suplatast tosilate group experienced rebound. There was no enhancement of production of immunoglobulins and cytokines after 2 weeks of treatment. Enhanced production of the Th2-cell cytokines, which selectively induces IgE and IgG4 production, may be involved in the pathogenesis of the rebound phenomenon, and that suplatast tosilate may prevent the rebound phenomenon by down-regulating the production of these cytokines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0760
Bufexamac, analytical standard
C12H17NO3