EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Regioselective hydrolysis of peracetylated alpha-D-glycopyranose catalyzed by immobilized lipases in aqueous medium. A facile preparation of useful intermediates for oligosaccharide synthesis.
PMID 10098679
Abstract
Penta-O-acetyl-alpha-D-glucopyranose was selectively deacetylated in aqueous media by lipases from Candida cilindracea (CCL) adsorbed on octyl-agarose support. Enzymatic hydrolyses was regioselective at the 4-position under neutral pH and towards the 6 position under acidic conditions. This enzymatic approach allows the one step synthesis of 1,2,3,6-tetra-O-acetyl-alpha-D-glucopyranoses 1, a useful intermediate in oligosaccharide synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2354
α-D(+)-Glucose pentaacetate, 99%
C16H22O11
285943
β-D-Glucose pentaacetate, 98%
C16H22O11