EMAIL THIS PAGE TO A FRIEND
Chemosphere
Identification of the major urinary and fecal metabolites of 3,5-dinitrobenzamide in chickens and rats.
PMID 10101847
Abstract
Colostomized chickens given oral doses of 3,5-dinitrobenzamide (nitromide) cleared nitromide predominantly through the urine (58% of dose) and feces (21% of dose). Rats cleared 52% of nitromide via urinary excretion and 44% via feces. Major urinary metabolites for both chickens and rats include: 3-amino-5-nitrobenzamide, 3-acetamido-5-nitrobenzamide, 3-acetamide-5-aminobenzamide, and 3,5-diacetamidobenzamide. The major fecal metabolite in chickens was 3-acetamido-5-nitrobenzamide (67% of fecal 14C) and 3-acetamido-5-aminobenzamide in rats (approximately 50%).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D193801
3,5-Dinitrobenzamide, 97%
C7H5N3O5
46518
Nitromide, VETRANAL, analytical standard
C7H5N3O5