EMAIL THIS PAGE TO A FRIEND
Clinical materials
Biological performance of tantalum.
PMID 10172264
Abstract
A detailed literature search was carried out to define the current knowledge about the biological performance of tantalum. The pure metal appears, to a great degree, to be inert both in vivo and in vitro. Both the pure metal and its principle oxide possess low solubility and toxicity; however, halide compounds are more biologically active. Local host response is benign, characterized by vital encapsulation in soft tissue and frequent osteointegration, reminiscent of titanium, in hard tissue. Tantalum has been in clinical use since before 1940 and has found a wide range of diagnostic and implant applications, with apparently overall excellent results. In some applications, such as for radiographic bone markers and cranial closure, tantalum may well be the current material of choice. In summary, metallic tantalum is a promising biomaterial whose applications have been limited by technical rather than biological performance considerations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
262889
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Ta
357251
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Ta
262897
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Ta
357243
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Ta
262919
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Ta
545007
Tantalum, powder, 99.99% trace metals basis
Ta
692824
Tantalum, powder, 60-100 mesh, 99.9%
Ta
262846
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Ta
593486
Tantalum, nanopowder, <25 nm particle size (BET), ≥99% trace metals basis
Ta
262854
Tantalum, rod, diam. 6.35 mm, ≥99.9% trace metals basis
Ta
356980
Tantalum, rod, diam. 3.2 mm, ≥99.9% trace metals basis
Ta
262862
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Ta
357006
Tantalum, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Ta
767514
Tantalum, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Ta
GF34465888
Tantalum, foil, 0.5m coil, thickness 0.034mm, as rolled, 99.9%
Ta
GF96084603
Tantalum, foil, 0.5m coil, thickness 0.03mm, as rolled, 99.9%
Ta
GF78648092
Tantalum, foil, 0.5m coil, thickness 0.03mm, coil width 1.2mm, annealed, 99.9%
Ta
GF38365106
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76540181
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Ta
GF08928108
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 1mm, annealed, 99.9%
Ta
GF61023574
Tantalum, foil, 0.5m coil, thickness 0.05mm, annealed, 99.9%
Ta
GF07210347
Tantalum, foil, 10m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Ta
GF10417698
Tantalum, foil, 10m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%
Ta
GF53229269
Tantalum, foil, 10m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76534500
Tantalum, foil, 100x100mm, thickness 0.100mm, annealed, 99.9%
Ta
GF80276182
Tantalum, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Ta
GF85784653
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, 99.9%
Ta
GF65422739
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, flattened, 99.9%
Ta
GF86342485
Tantalum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Ta
GF60298195
Tantalum, foil, 100x100mm, thickness 0.25mm, annealed, 99.9%
Ta