EMAIL THIS PAGE TO A FRIEND
Biotechnic & histochemistry : official publication of the Biological Stain Commission
Chemical dehydration of specimens with 2,2-dimethoxypropane (DMP) for paraffin processing of animal tissues: practical and economic advantages over dehydration in ethanol.
PMID 10190257
Abstract
Chemical dehydration can be accomplished using 2,2-dimethoxypropane (DMP). In the presence of an acid catalyst, this liquid reacts with water generating methanol and acetone as products. Although DMP is more expensive per milliliter than ethanol and other solvents used for dehydration, it is an economical alternative because a much smaller volume is needed. Slow penetration of DMP was previously thought to restrict its use to tiny specimens, but we now show that pieces of tissue as thick as 2 cm are dehydrated by overnight immersion in acidified DMP. We also show that dehydration in acidified DMP does not impair the staining of RNA or other basophilic components of animal tissues. The temperature and concentrations of methanol and H+ in the chemical dehydrating agent are too low to produce histochemically detectable methylation or nucleic acid extraction.