EMAIL THIS PAGE TO A FRIEND
Experimental neurology
Isolation and cloning of multipotential stem cells from the embryonic human CNS and establishment of transplantable human neural stem cell lines by epigenetic stimulation.
PMID 10192778
Abstract
Stem cells that can give rise to neurons, astroglia, and oligodendroglia have been found in the developing and adult central nervous system (CNS) of rodents. Yet, their existence within the human brain has not been documented, and the isolation and characterization of multipotent embryonic human neural stem cells have proven difficult to accomplish. We show that the developing human CNS embodies multipotent precursors that differ from their murine counterpart in that they require simultaneous, synergistic stimulation by both epidermal and fibroblast growth factor-2 to exhibit critical stem cell characteristics. Clonal analysis demonstrates that human C NS stem cells are multipotent and differentiate spontaneously into neurons, astrocytes, and oligodendrocytes when growth factors are removed. Subcloning and population analysis show their extensive self-renewal capacity and functional stability, their ability to maintain a steady growth profile, their multipotency, and a constant potential for neuronal differentiation for more than 2 years. The neurons generated by human stem cells over this period of time are electrophysiologically active. These cells are also cryopreservable. Finally, we demonstrate that the neuronal and glial progeny of long-term cultured human CNS stem cells can effectively survive transplantation into the lesioned striatum of adult rats. Tumor formation is not observed, even in immunodeficient hosts. Hence, as a consequence of their inherent biology, human CNS stem cells can establish stable, transplantable cell lines by epigenetic stimulation. These lines represent a renewable source of neurons and glia and may significantly facilitate research on human neurogenesis and the development of clinical neural transplantation.