EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Characterization of the binding site of the histamine H3 receptor. 1. Various approaches to the synthesis of 2-(1H-imidazol-4-yl)cyclopropylamine and histaminergic activity of (1R,2R)- and (1S,2S)-2-(1H-imidazol-4-yl)-cyclopropylamine.
PMID 10197956
Abstract
Various approaches to the synthesis of all four stereoisomers of 2-(1H-imidazol-4-yl)cyclopropylamine (cyclopropylhistamine) are described. The rapid and convenient synthesis and resolution of trans-cyclopropylhistamine is reported. The absolute configuration of its enantiomers was determined by single-crystal X-ray crystallographic analysis. The distinct trans-cyclopropylhistamine enantiomers were tested for their activity and affinity on the histamine H3 receptor. (1S,2S)-Cyclopropylhistamine (VUF 5297) acts as an agonist both on the rat cortex (pD2 = 7.1; alpha = 0.75) and on guinea pig jejunum (pD2 = 6.6; alpha = 0.75). Its enantiomer, (1R, 2R)-cyclopropylhistamine (VUF 5296), is about 1 order of magnitude less active. Both enantiomers show weak activity on H1 and H2 receptors. All synthetic attempts to cis-cyclopropylhistamine were unsuccessful. Nevertheless, the results of this study provide an ideal template for molecular modeling studies of histamine H3 receptor ligands.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
157279
Ethyl 2-formyl-1-cyclopropanecarboxylate, predominantly trans, 96%
C7H10O3