EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
In vitro synthesis of peptidoglycan precursors modified with N-acetylputrescine by Cyanophora paradoxa cyanelle envelope membranes.
PMID 10198034
Abstract
The photosynthetic organelles (cyanelles) of the protist Cyanophora paradoxa are surrounded by a peptidoglycan wall, modified through amidation with N-acetylputrescine. Cyanelle envelope membrane preparations were shown to catalyze the lipid-linked steps of peptidoglycan biosynthesis as well as the putrescinylation and subsequent acetylation, occurring at the stage of lipid I and/or lipid II.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8784
N-Acetylputrescine hydrochloride, ≥98% (TLC)
C6H14N2O · HCl