EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[The possibility of using risk assessments of the development of diagnostically different body states for setting hygienic standards].
PMID 10199093
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A10000
Acetone cyanohydrin, 99%
C4H7NO