EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Femtochemistry uncovers the nature of electron transfer reactions.
PMID 10200240
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
107247
Diethyl sulfide, 98%
C4H10S
W382501
Diethyl sulfide, 98%, FG
C4H10S