EMAIL THIS PAGE TO A FRIEND
Anesthesiology
Anaphylactic shock to neuromuscular blocking agent: a familial history.
PMID 10201697
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0325200 Alcuronium chloride, European Pharmacopoeia (EP) Reference Standard
C44H50Cl2N4O2