EMAIL THIS PAGE TO A FRIEND
Pediatric research
Profile of urinary bile acids in infants and children: developmental pattern of excretion of unsaturated ketonic bile acids and 7beta-hydroxylated bile acids.
PMID 10203155
Abstract
Unusual bile acids, such as unsaturated ketonic and 7beta-hydroxylated bile acids, have been detected in urine early in life. To elucidate the normal profiles of usual and unusual urinary bile acids in the neonatal and pediatric periods, we measured the concentrations of 28 kinds in urine from normal newborns, infants, and children by gas chromatography-mass spectrometry. The mean total bile acid/Cr ratio in 7-d-old infants was significantly higher than in subjects of other age groups (birth, 2-4 mo, 5-7 mo, 11-12 mo, 2-3 y, 9-14 y, and adult) (p < 0.05). Relatively large amounts of unusual bile acids were detected during infancy, especially during the period up to 1 mo of age. At that time, 1beta,3alpha,7alpha,12alpha-tetrahydroxy-5bet a-cholan-24-oic, 7alpha, 12alpha-dihydroxy-3-oxo-5beta-chol-1-en-24-oic, and 7alpha,12alpha-dihydroxy-3-oxo-4-cholen-24-oic acids were predominant among the unusual urinary bile acids present. Moreover, the levels of 3alpha,7beta,12alpha-trihydroxy-5beta-cholan+ ++-24-oic acid increased significantly after 2-4 mo of age. These results indicate that bile acid synthesis and metabolism in the liver of developing infants are significantly different from that occurring in the liver of adults. Significant amounts of urinary isomerized 7beta-hydroxylated bile acids were detected after late infancy, probably because of changes in the intestinal bacterial flora response to a change in nutrition. We describe, for the first time, evidence of the epimerization of the 7alpha-hydroxyl group of cholic acid, which may be unique to human development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
41605
DL-Mandelic acid, analytical standard
C8H8O3
M2101
Mandelic acid, 99%
C8H8O3