EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Polymer-bound N-hydroxysuccinimide esters: a column-free fluorescent-labeling method.
PMID 10206554
Abstract
Polymer-bound N-hydroxysuccinimide esters of 1-pyrenebutyric acid, 6-carboxyfluorescein diacetate, and biotin were efficiently prepared. Column-free fluorescent- and biotin-labeling reactions of various amines using these resins were successfully demonstrated.