EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Inhibition of T cell signaling by mitogen-activated protein kinase-targeted hematopoietic tyrosine phosphatase (HePTP).
PMID 10206983
Abstract
Activation of T lymphocytes to produce cytokines is regulated by the counterbalance of protein-tyrosine kinases and protein-tyrosine phosphatases, many of which have a high degree of substrate specificity because of physical association with their targets. Overexpression of hematopoietic protein-tyrosine phosphatase (HePTP) results in suppression of T lymphocyte activation as measured by T cell antigen receptor-induced activation of transcription factors binding to the 5' promoter of the interleukin-2 gene. Efforts to pinpoint the exact site of action and specificity of HePTP in the signaling cascade revealed that HePTP acts directly on the mitogen-activated protein (MAP) kinases Erk1 and 2 and consequently reduces the magnitude and duration of their catalytic activation in intact T cells. In contrast, HePTP had no effects on N-terminal c-Jun kinase or on events upstream of the MAP kinases. The specificity of HePTP correlated with its physical association through its noncatalytic N terminus with Erk and another MAP kinase, p38, but not Jnk or other proteins. We propose that HePTP plays a negative role in antigen receptor signaling by specifically regulating MAP kinases in the cytosol and at early time points of T cell activation before the activation-induced expression of nuclear dual-specific MAP kinase phosphatases.