EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Merbromine-induced pustular contact dermatitis.
PMID 10208516
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7011
Mercury dibromofluorescein disodium salt, practical grade, crystalline
C20H8Br2HgNa2O6