EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Allergic contact dermatitis associated with photosensitivity, from alantolactone in a chrysanthemum farmer.
PMID 10208523
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0415
Alantolactone, ≥98% (HPLC)
C15H20O2