EMAIL THIS PAGE TO A FRIEND
Developmental medicine and child neurology
Pharmacological management of convulsive status epilepticus in children.
PMID 10210254
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5520
Paraldehyde, ≥97%
C6H12O3