EMAIL THIS PAGE TO A FRIEND
Experimental neurology
Creatine and cyclocreatine attenuate MPTP neurotoxicity.
PMID 10222117
Abstract
Systemic administration of 1-methyl-4-phenyl-1,2,3, 6-tetrahydropyridine (MPTP) produces parkinsonism in experimental animals by a mechanism involving impaired energy production. MPTP is converted by monoamine oxidase B to 1-methyl-4-phenylpyridinium (MPP+), which blocks complex I of the electron transport chain. Oral supplementation with creatine or cyclocreatine, which are substrates for creatine kinase, may increase phosphocreatine (PCr) or cyclophosphocreatine (PCCr) and buffer against ATP depletion and thereby exert neuroprotective effects. In the present study we found that oral supplementation with either creatine or cyclocreatine produced significant protection against MPTP-induced dopamine depletions in mice. Creatine protected against MPTP-induced loss of Nissl and tyrosine hydroxylase immunostained neurons in the substantia nigra. Creatine and cyclocreatine had no effects on the conversion of MPTP to MPP+ in vivo. These results further implicate metabolic dysfunction in MPTP neurotoxicity and suggest a novel therapeutic approach, which may have applicability for Parkinson's disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
377627
2-Imino-1-imidazolidineacetic acid, 98%
C5H9N3O2